Tisk…

3-Fluoro-4-methoxybenzamide 97%

Dodavatel: Apollo Scientific
Varování

PC302054-25G PC302054-5G PC302054-1G PC302054-100G
APOSPC302054-25GEA 6390 CZK
APOSPC302054-25G APOSPC302054-5G APOSPC302054-1G APOSPC302054-100G
3-Fluoro-4-methoxybenzamide 97%
3-Fluoro-4-methoxybenzamide
Vzorec: C₈H₈FNO₂
MW: 169,153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660163
CAS číslo: 701640-04-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR