2-Methoxy-4-(trifluoromethoxy)phenylacetonitrile 97%
Dodavatel: Apollo Scientific
PC302028-1G
PC302028-5G
PC302028-10G
APOSPC302028-1GEA
3440
CZK
APOSPC302028-1G
APOSPC302028-5G
APOSPC302028-10G
2-Methoxy-4-(trifluoromethoxy)phenylacetonitrile 97%
Vzorec:
C₁₀H₈F₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD04115971 CAS číslo: 886500-40-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...