Tisk…

2-Fluoro-5-methylcinnamic acid 98%

Dodavatel: Apollo Scientific

PC302027-1G PC302027-5G
APOSPC302027-1GEA 2980 CZK
APOSPC302027-1G APOSPC302027-5G
2-Fluoro-5-methylcinnamic acid 98%
2-Fluoro-5-methylcinnamic acid
Vzorec: C₁₀H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD06205468
CAS číslo: 773130-21-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR