Tisk…

2-Chloro-4,5-difluorobenzamide 98%

Dodavatel: Apollo Scientific

PC302021-5G PC302021-1G
APOSPC302021-5GEA 7250 CZK
APOSPC302021-5G APOSPC302021-1G
2-Chloro-4,5-difluorobenzamide 98%
2-Chloro-4,5-difluorobenzamide
Vzorec: C₇H₄ClF₂NO
MW: 191,563 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02628151
CAS číslo: 296274-32-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR