2-[2-Fluoro-3-(trifluoromethyl)phenyl]ethanol 95%
Dodavatel: Apollo Scientific
PC302017-1G
PC302017-5G
APOSPC302017-1GEA
4100
CZK
APOSPC302017-1G
APOSPC302017-5G
2-[2-Fluoro-3-(trifluoromethyl)phenyl]ethanol 95%
2-[2-Fluoro-3-(trifluoromethyl)phenyl]ethanol
Vzorec:
C₉H₈F₄O MW: 208,153 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06660360 CAS číslo: 886761-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...