Tisk…

2-(3-Chloro-2-fluorophenyl)ethanol 95%

Dodavatel: Apollo Scientific
Varování

PC302004-1G PC302004-5G
APOSPC302004-1GEA 11910 CZK
APOSPC302004-1G APOSPC302004-5G
2-(3-Chloro-2-fluorophenyl)ethanol 95%
2-(3-Chloro-2-fluorophenyl)ethanol
Vzorec: C₈H₈ClFO
MW: 174,6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660361
CAS číslo: 886761-82-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR