Tisk…
Ethyl 8-methyl-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Dodavatel: Apollo Scientific
PC301031-1G PC301031-250MG
APOSPC301031-1GEA 14880 CZK
APOSPC301031-1G APOSPC301031-250MG
Ethyl 8-methyl-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Ethyl 8-methyl-2-(difluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Vzorec: C₁₂H₁₂F₂N₂O₂
MW: 254.233 g/mol
Storage Temperature: Ambient
CAS číslo: 2514942-05-1