Tisk…

3,5-Difluorophenyl acetate

Dodavatel: Apollo Scientific

PC301027-1G PC301027-5G
APOSPC301027-1GEA 5630 CZK
APOSPC301027-1G APOSPC301027-5G
3,5-Difluorophenyl acetate
3,5-Difluorophenyl acetate
Vzorec: C₈H₆F₂O₂
MW: 172.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25977412
CAS číslo: 315690-94-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR