1-(4-Nitrophenyl)-4-[5-(trifluoromethyl)pyridin-2-yl]piperazine
Dodavatel: Apollo Scientific
PC300991-1G
PC300991-250MG
APOSPC300991-1GEA
12200
CZK
APOSPC300991-1G
APOSPC300991-250MG
1-(4-Nitrophenyl)-4-[5-(trifluoromethyl)pyridin-2-yl]piperazine
1-(4-Nitrophenyl)-4-[5-(trifluoromethyl)pyridin-2-yl]piperazine
Vzorec:
C₁₆H₁₅F₃N₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD31630107 CAS číslo: 1294374-23-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...