5-(Trifluoromethoxy)-1H-indole-2-carbohydrazide
Dodavatel: Apollo Scientific
|
Varování
|
PC300989-1G
PC300989-250MG
APOSPC300989-1GEA
53560
CZK
APOSPC300989-1G
APOSPC300989-250MG
5-(Trifluoromethoxy)-1H-indole-2-carbohydrazide
5-(Trifluoromethoxy)-1H-indole-2-carbohydrazide
Vzorec:
C₁₀H₈F₃N₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD04110840 CAS číslo: 685120-14-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...