Tisk…

Ethyl 2-bromo-4-fluorobenzoate

Dodavatel: Apollo Scientific
Varování

PC300979-1G PC300979-5G
APOSPC300979-1GEA 2480 CZK
APOSPC300979-1G APOSPC300979-5G
Ethyl 2-bromo-4-fluorobenzoate
Ethyl 2-bromo-4-fluorobenzoate
Vzorec: C₉H₈BrFO₂
MW: 247,061 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204924
CAS číslo: 651341-68-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR