(2Z)-4-[(3-Fluoro-4-methylphenyl)amino]-4-oxobut-2-enoic acid
Dodavatel: Apollo Scientific
PC300960-10G
PC300960-5G
PC300960-1G
APOSPC300960-10GEA
18520
CZK
APOSPC300960-10G
APOSPC300960-5G
APOSPC300960-1G
(2Z)-4-[(3-Fluoro-4-methylphenyl)amino]-4-oxobut-2-enoic acid
(2Z)-4-[(3-Fluoro-4-methylphenyl)amino]-4-oxobut-2-enoic acid
Vzorec:
C₁₁H₁₀FNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00452550 CAS číslo: 1421751-34-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...