Tisk…

2,4,5-Trifluorobenzohydrazide

Dodavatel: Apollo Scientific
Varování

PC300958-10G PC300958-1G PC300958-5G
APOSPC300958-10GEA 9530 CZK
APOSPC300958-10G APOSPC300958-1G APOSPC300958-5G
2,4,5-Trifluorobenzohydrazide
2,4,5-Trifluorobenzohydrazide
Vzorec: C₇H₅F₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD22413548
CAS číslo: 1824155-80-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR