2,2,2-Trifluoro-1-(1-methyl-1H-pyrrol-2-yl)ethanone
Dodavatel: Apollo Scientific
PC300942-10G
PC300942-5G
PC300942-1G
APOSPC300942-10GEA
14060
CZK
APOSPC300942-10G
APOSPC300942-5G
APOSPC300942-1G
2,2,2-Trifluoro-1-(1-methyl-1H-pyrrol-2-yl)ethanone
2,2,2-Trifluoro-1-(1-methyl-1H-pyrrol-2-yl)ethanone
Vzorec:
C₇H₆F₃NO MW: 177,124 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07852052 CAS číslo: 18087-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...