Tisk…

1-(3-Chloro-4-fluorophenyl)-3-(dimethylamino)prop-2-en-1-one

Dodavatel: Apollo Scientific
Varování

PC300889-1G PC300889-5G
APOSPC300889-1GEA 992 CZK
APOSPC300889-1G APOSPC300889-5G
1-(3-Chloro-4-fluorophenyl)-3-(dimethylamino)prop-2-en-1-one
1-(3-Chloro-4-fluorophenyl)-3-(dimethylamino)prop-2-en-1-one
Vzorec: C₁₁H₁₁ClFNO
MW: 227,663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01570079
CAS číslo: 154258-48-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR