N,N-Diisopropyl-4-(trifluoromethoxy)benzenesulphonamide
Dodavatel: Apollo Scientific
PC300883-10G
PC300883-1G
PC300883-5G
APOSPC300883-10GEA
24730
CZK
APOSPC300883-10G
APOSPC300883-1G
APOSPC300883-5G
N,N-Diisopropyl-4-(trifluoromethoxy)benzenesulphonamide
N,N-Diisopropyl-4-(trifluoromethoxy)benzenesulphonamide
Vzorec:
C₁₃H₁₈F₃NO₃S MW: 325.347 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28992264 CAS číslo: 1993322-94-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...