Tisk…

1,1,1-Trifluoro-3-piperidin-1-ylpropan-2-ol

Dodavatel: Apollo Scientific
Varování

PC300875-1G PC300875-250MG PC300875-5G
APOSPC300875-1GEA 827 CZK
APOSPC300875-1G APOSPC300875-250MG APOSPC300875-5G
1,1,1-Trifluoro-3-piperidin-1-ylpropan-2-ol
1,1,1-Trifluoro-3-piperidin-1-ylpropan-2-ol
Vzorec: C₈H₁₄F₃NO
Storage Temperature: Ambient
MDL Number: MFCD00551090
CAS číslo: 478050-22-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR