Tisk…

1,1,1-Trifluoro-3-[4-(2-hydroxyethyl)piperazin-1-yl]propan-2-ol

Dodavatel: Apollo Scientific
Varování

PC300871-1G PC300871-250MG PC300871-5G
APOSPC300871-1GEA 1990 CZK
APOSPC300871-1G APOSPC300871-250MG APOSPC300871-5G
1,1,1-Trifluoro-3-[4-(2-hydroxyethyl)piperazin-1-yl]propan-2-ol
1,1,1-Trifluoro-3-[4-(2-hydroxyethyl)piperazin-1-yl]propan-2-ol
Vzorec: C₉H₁₇F₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD02082049
CAS číslo: 477847-03-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR