Tisk…

Ethyl chloro[(2-fluorophenyl)hydrazono]acetate

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Chloro-2-(2-fluoro-phenyl-hydrazono)-acetic acid ethyl ester

PC300846-1G PC300846-5G
APOSPC300846-1GEA 4370 CZK
APOSPC300846-1G APOSPC300846-5G
Ethyl chloro[(2-fluorophenyl)hydrazono]acetate
Ethyl chloro[(2-fluorophenyl)hydrazono]acetate
Vzorec: C₁₀H₁₀ClFN₂O₂
MW: 244,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06809224
CAS číslo: 64989-74-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR