N-(4-Chlorophenyl)-N'-[4-(trifluoromethoxy)phenyl]urea
Dodavatel: Apollo Scientific
PC300822-1G
PC300822-5G
APOSPC300822-1GEA
3540
CZK
APOSPC300822-1G
APOSPC300822-5G
N-(4-Chlorophenyl)-N'-[4-(trifluoromethoxy)phenyl]urea
N-(4-Chlorophenyl)-N'-[4-(trifluoromethoxy)phenyl]urea
Vzorec:
C₁₄H₁₀ClF₃N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD02632654 CAS číslo: 54730-71-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...