Tisk…

Methyl 2,5-difluoro-4-piperidin-1-ylbenzoate

Dodavatel: Apollo Scientific

PC300791-1G PC300791-250MG PC300791-5G
APOSPC300791-1GEA 8270 CZK
APOSPC300791-1G APOSPC300791-250MG APOSPC300791-5G
Methyl 2,5-difluoro-4-piperidin-1-ylbenzoate
Methyl 2,5-difluoro-4-piperidin-1-ylbenzoate
Vzorec: C₁₃H₁₅F₂NO₂
MW: 255,261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103352
CAS číslo: 1858241-61-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR