Tisk…

Methyl 2,5-difluoro-4-morpholin-4-ylbenzoate

Dodavatel: Apollo Scientific

PC300786-1G PC300786-250MG
APOSPC300786-1GEA 8270 CZK
APOSPC300786-1G APOSPC300786-250MG
Methyl 2,5-difluoro-4-morpholin-4-ylbenzoate
Methyl 2,5-difluoro-4-morpholin-4-ylbenzoate
Vzorec: C₁₂H₁₃F₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD28103347
CAS číslo: 1858251-27-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR