Tisk…

4-(Trifluoromethyl)pyrimidine-5-carboxylic acid

Dodavatel: Apollo Scientific

PC300766-1G PC300766-250MG
APOSPC300766-1GEA 3150 CZK
APOSPC300766-1G APOSPC300766-250MG
4-(Trifluoromethyl)pyrimidine-5-carboxylic acid
4-(Trifluoromethyl)pyrimidine-5-carboxylic acid
Vzorec: C₆H₃F₃N₂O₂
MW: 192,095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11106713
CAS číslo: 220880-12-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR