Tisk…

3-{4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]homopiperazin-1-yl}propanoic acid

Dodavatel: Apollo Scientific

Synonyms: 3-{4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1,4-diazepan-1-yl}propanoic acid

PC300724-1G PC300724-10G PC300724-5G
APOSPC300724-1GEA 9590 CZK
APOSPC300724-1G APOSPC300724-10G APOSPC300724-5G
3-{4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]homopiperazin-1-yl}propanoic acid
3-{4-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]homopiperazin-1-yl}propanoic acid
Vzorec: C₁₄H₁₇ClF₃N₃O₂
MW: 351,752 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12756237
CAS číslo: 1227954-72-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR