Tisk…

3-{4-[5-(Trifluoromethyl)pyridin-2-yl]homopiperazin-1-yl}propanoic acid

Dodavatel: Apollo Scientific

Synonyms: 3-{4-[5-(Trifluoromethyl)pyridin-2-yl]-1,4-diazepan-1-yl}propanoic acid

PC300722-1G PC300722-10G PC300722-5G
APOSPC300722-1GEA 9590 CZK
APOSPC300722-1G APOSPC300722-10G APOSPC300722-5G
3-{4-[5-(Trifluoromethyl)pyridin-2-yl]homopiperazin-1-yl}propanoic acid
3-{4-[5-(Trifluoromethyl)pyridin-2-yl]homopiperazin-1-yl}propanoic acid
Vzorec: C₁₄H₁₈F₃N₃O₂
MW: 317,307 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12545992
CAS číslo: 1227954-71-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR