Tisk…

Ethyl 5-chloro-2-fluorobenzoate

Dodavatel: Apollo Scientific
Varování

PC300718-1G PC300718-5G
APOSPC300718-1GEA 2780 CZK
APOSPC300718-1G APOSPC300718-5G
Ethyl 5-chloro-2-fluorobenzoate
Ethyl 5-chloro-2-fluorobenzoate
Vzorec: C₉H₈ClFO₂
MW: 202,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06205125
CAS číslo: 773139-56-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR