Tisk…

(2Z)-4-{[3-(Difluoromethoxy)phenyl]amino}-4-oxobut-2-enoic acid

Dodavatel: Apollo Scientific

PC300699-1G PC300699-250MG PC300699-5G
APOSPC300699-1GEA 4630 CZK
APOSPC300699-1G APOSPC300699-250MG APOSPC300699-5G
(2Z)-4-{[3-(Difluoromethoxy)phenyl]amino}-4-oxobut-2-enoic acid
(2Z)-4-{[3-(Difluoromethoxy)phenyl]amino}-4-oxobut-2-enoic acid
Vzorec: C₁₁H₉F₂NO₄
MW: 257,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20037424
CAS číslo: 1216321-25-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR