Methyl 2,4,5-trifluorobenzoate 99%
Dodavatel: Apollo Scientific
|
Varování
|
PC300696-100G
PC300696-10G
PC300696-25G
APOSPC300696-100GEA
8270
CZK
APOSPC300696-100G
APOSPC300696-10G
APOSPC300696-25G
Methyl 2,4,5-trifluorobenzoate 99%
Methyl 2,4,5-trifluorobenzoate
Vzorec:
C₈H₅F₃O₂ MW: 190,119 g/mol Bod varu: 130…135 °C (100 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD06203702 CAS číslo: 20372-66-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...