1-[5-(Trifluoromethyl)pyridin-2-yl]homopiperazine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1-(5-(Trifluoromethyl)pyridin-2-yl)-1,4-diazepane, 1-[5-(Trifluoromethyl)pyridin-2-yl]-1,4-diazepane
PC300684-10G
PC300684-1G
PC300684-5G
APOSPC300684-10GEA
10950
CZK
APOSPC300684-10G
APOSPC300684-1G
APOSPC300684-5G
1-[5-(Trifluoromethyl)pyridin-2-yl]homopiperazine
1-[5-(Trifluoromethyl)pyridin-2-yl]homopiperazine
Vzorec:
C₁₁H₁₄F₃N₃ MW: 245.244 g/mol Bod varu: 110 °C (0.1 mm Hg) Bod tání: 50…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00661783 CAS číslo: 306934-70-3 UN: 3259 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...