Tisk…

Methyl 2-amino-6-[4-(trifluoromethyl)pyrimidin-2-yl]-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate

Dodavatel: Apollo Scientific

PC300677-1G PC300677-5G
APOSPC300677-1GEA 8440 CZK
APOSPC300677-1G APOSPC300677-5G
Methyl 2-amino-6-[4-(trifluoromethyl)pyrimidin-2-yl]-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate
Methyl 2-amino-6-[4-(trifluoromethyl)pyrimidin-2-yl]-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylate
Vzorec: C₁₄H₁₃F₃N₄O₂S
MW: 358,339 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16556315
CAS číslo: 1427460-39-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR