Tisk…

1,3-Dimethyl-5-(trifluoromethyl)pyrido[2,3-d]pyrimidine-2,4,7(1H,3H,8H)-trione

Dodavatel: Apollo Scientific

PC300671-1G PC300671-10G PC300671-5G
APOSPC300671-1GEA 5630 CZK
APOSPC300671-1G APOSPC300671-10G APOSPC300671-5G
1,3-Dimethyl-5-(trifluoromethyl)pyrido[2,3-d]pyrimidine-2,4,7(1H,3H,8H)-trione
1,3-Dimethyl-5-(trifluoromethyl)pyrido[2,3-d]pyrimidine-2,4,7(1H,3H,8H)-trione
Vzorec: C₁₀H₈F₃N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD23159171
CAS číslo: 779352-01-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR