Ethyl 1-[1,3-dimethyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-6-yl]piperidine-4-carboxylate
Dodavatel: Apollo Scientific
PC300668-1G
PC300668-10G
PC300668-5G
APOSPC300668-1GEA
7740
CZK
APOSPC300668-1G
APOSPC300668-10G
APOSPC300668-5G
Ethyl 1-[1,3-dimethyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-6-yl]piperidine-4-carboxylate
Ethyl 1-[1,3-dimethyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-6-yl]piperidine-4-carboxylate
Vzorec:
C₁₇H₂₁F₃N₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD13194593 CAS číslo: 1305712-96-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...