Tisk…

2-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzaldehyde

Dodavatel: Apollo Scientific

PC300648-1G PC300648-10G PC300648-5G
APOSPC300648-1GEA 5490 CZK
APOSPC300648-1G APOSPC300648-10G APOSPC300648-5G
2-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzaldehyde
2-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}benzaldehyde
Vzorec: C₁₂H₇F₃N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11846261
CAS číslo: 1227954-93-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR