Tisk…

1-(3-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}phenyl)ethanone

Dodavatel: Apollo Scientific

PC300629-1G PC300629-5G
APOSPC300629-1GEA 7050 CZK
APOSPC300629-1G APOSPC300629-5G
1-(3-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}phenyl)ethanone
1-(3-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}phenyl)ethanone
Vzorec: C₁₄H₁₀F₃NO₂
MW: 281,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22370025
CAS číslo: 1427460-55-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR