Tisk…

(4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}phenyl)methanol

Dodavatel: Apollo Scientific

PC300594-1G PC300594-5G
APOSPC300594-1GEA 4240 CZK
APOSPC300594-1G APOSPC300594-5G
(4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}phenyl)methanol
(4-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}phenyl)methanol
Vzorec: C₁₃H₉ClF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD12756229
CAS číslo: 1160430-73-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR