2-Bromo-6-fluorobenzyl alcohol
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(2-Bromo-6-fluorophenyl)methanol
PC300574-25G
PC300574-5G
APOSPC300574-25GEA
2820
CZK
APOSPC300574-25G
APOSPC300574-5G
2-Bromo-6-fluorobenzyl alcohol
2-Bromo-6-fluorobenzyl alcohol
Vzorec:
C₇H₆BrFO MW: 205.024 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09263985 CAS číslo: 261723-33-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...