7-Fluoro-6-methoxy-1H-indazol-3-amine
Dodavatel: Apollo Scientific
|
Varování
|
PC300572-1G
PC300572-250MG
APOSPC300572-1GEA
7770
CZK
APOSPC300572-1G
APOSPC300572-250MG
7-Fluoro-6-methoxy-1H-indazol-3-amine
7-Fluoro-6-methoxy-1H-indazol-3-amine
Vzorec:
C₈H₈FN₃O MW: 181.167 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16631265 CAS číslo: 1279842-87-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...