2-(4-Bromo-1H-pyrazol-1-yl)-3-chloro-5-(trifluoromethyl)pyridine
Dodavatel: Apollo Scientific
PC300537-1G
PC300537-5G
PC300537-10G
APOSPC300537-1GEA
5290
CZK
APOSPC300537-1G
APOSPC300537-5G
APOSPC300537-10G
2-(4-Bromo-1H-pyrazol-1-yl)-3-chloro-5-(trifluoromethyl)pyridine
2-(4-Bromo-1H-pyrazol-1-yl)-3-chloro-5-(trifluoromethyl)pyridine
Vzorec:
C₉H₄BrClF₃N₃ MW: 326,5 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12861968 CAS číslo: 1183812-46-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...