2-Methyl-6-(trifluoromethyl)pyridine-3-sulphonamide
Dodavatel: Apollo Scientific
|
Varování
|
PC300533-1G
PC300533-100MG
PC300533-250MG
APOSPC300533-1GEA
0
CZK
APOSPC300533-1G
APOSPC300533-100MG
APOSPC300533-250MG
2-Methyl-6-(trifluoromethyl)pyridine-3-sulphonamide
2-Methyl-6-(trifluoromethyl)pyridine-3-sulphonamide
Vzorec:
C₇H₇F₃N₂O₂S MW: 240,203 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22577294 CAS číslo: 1427461-10-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...