Tisk…

4-(4-Fluorophenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one

Dodavatel: Apollo Scientific
Varování

PC300521-1G PC300521-5G
APOSPC300521-1GEA 1990 CZK
APOSPC300521-1G APOSPC300521-5G
4-(4-Fluorophenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one
4-(4-Fluorophenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one
Vzorec: C₈H₆FN₃O
MW: 179,151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11053874
CAS číslo: 80240-40-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR