[2-Morpholin-4-yl-5-(trifluoromethyl)phenyl]methanol
Dodavatel: Apollo Scientific
|
Varování
|
PC300518-1G
PC300518-250MG
APOSPC300518-1GEA
2900
CZK
APOSPC300518-1G
APOSPC300518-250MG
[2-Morpholin-4-yl-5-(trifluoromethyl)phenyl]methanol
[2-Morpholin-4-yl-5-(trifluoromethyl)phenyl]methanol
Vzorec:
C₁₂H₁₄F₃NO₂ MW: 261,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09025885 CAS číslo: 886851-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...