2-Bromo-7-fluorophenazine 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC29357-1G
PC29357-250MG
PC29357-5G
APOSPC29357-1GEA
5630
CZK
APOSPC29357-1G
APOSPC29357-250MG
APOSPC29357-5G
2-Bromo-7-fluorophenazine 95%
2-Bromo-7-fluorophenazine
Vzorec:
C₁₂H₆BrFN₂ MW: 277,092 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32066752 CAS číslo: 2366994-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...