7H-Perfluoro-4-methyl-3,6-dioxaoctanesulphonic acid
Dodavatel: Apollo Scientific
PC28311-1G
PC28311-250MG
APOSPC28311-1GEA
22120
CZK
APOSPC28311-1G
APOSPC28311-250MG
7H-Perfluoro-4-methyl-3,6-dioxaoctanesulphonic acid
7H-Perfluoro-4-methyl-3,6-dioxaoctanesulphonic acid
Vzorec:
C₇H₂F₁₄O₅S MW: 464,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32011184 CAS číslo: 749836-20-2 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...