Tisk…

Methyl 3-fluoro-5-(trifluoromethyl)picolinate

Dodavatel: Apollo Scientific
Varování

PC28303-1G PC28303-250MG
APOSPC28303-1GEA 19510 CZK
APOSPC28303-1G APOSPC28303-250MG
Methyl 3-fluoro-5-(trifluoromethyl)picolinate
Methyl 3-fluoro-5-(trifluoromethyl)picolinate
Vzorec: C₈H₅F₄NO₂
MW: 223.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28799119
CAS číslo: 1803834-87-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR