2-(Prop-2-yn-1-ylthio)-6-(trifluoromethyl)pyrimidin-4(3H)-one
Dodavatel: Apollo Scientific
|
Varování
|
PC28268-1G
PC28268-250MG
APOSPC28268-1GEA
6350
CZK
APOSPC28268-1G
APOSPC28268-250MG
2-(Prop-2-yn-1-ylthio)-6-(trifluoromethyl)pyrimidin-4(3H)-one
2-(Prop-2-yn-1-ylthio)-6-(trifluoromethyl)pyrimidin-4(3H)-one
Vzorec:
C₈H₅F₃N₂OS MW: 234,198 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08018587 CAS číslo: 141598-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...