4-(Trifluoromethyl)-1H-imidazole-2(3H)-thione
Dodavatel: Apollo Scientific
|
Varování
|
PC28263-1G
PC28263-250MG
APOSPC28263-1GEA
14920
CZK
APOSPC28263-1G
APOSPC28263-250MG
4-(Trifluoromethyl)-1H-imidazole-2(3H)-thione
4-(Trifluoromethyl)-1H-imidazole-2(3H)-thione
Vzorec:
C₄H₃F₃N₂S MW: 168,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28676943 CAS číslo: 149432-40-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...