Tisk…

4-Chloro-2-(methylsulphonyl)-6-(trifluoromethyl)pyrimidine

Dodavatel: Apollo Scientific
Varování

PC28262-1G PC28262-250MG
APOSPC28262-1GEA 16470 CZK
APOSPC28262-1G APOSPC28262-250MG
4-Chloro-2-(methylsulphonyl)-6-(trifluoromethyl)pyrimidine
4-Chloro-2-(methylsulphonyl)-6-(trifluoromethyl)pyrimidine
Vzorec: C₆H₄ClF₃N₂O₂S
MW: 260,621 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31616619
CAS číslo: 119610-21-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR