5-(Trifluoromethoxy)benzofuran-2-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC28256-1G
PC28256-250MG
APOSPC28256-1GEA
16370
CZK
APOSPC28256-1G
APOSPC28256-250MG
5-(Trifluoromethoxy)benzofuran-2-carboxylic acid
5-(Trifluoromethoxy)benzofuran-2-carboxylic acid
Vzorec:
C₁₀H₅F₃O₄ MW: 246,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD27925745 CAS číslo: 575469-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...