7-Bromo-5-(trifluoromethoxy)benzofuran-2-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC28250-1G
PC28250-250MG
APOSPC28250-1GEA
4770
CZK
APOSPC28250-1G
APOSPC28250-250MG
7-Bromo-5-(trifluoromethoxy)benzofuran-2-carboxylic acid
7-Bromo-5-(trifluoromethoxy)benzofuran-2-carboxylic acid
Vzorec:
C₁₀H₄BrF₃O₄ MW: 325.036 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31759996 CAS číslo: 2379918-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...