Tisk…

6-Fluoro-8-(trifluoromethyl)quinolin-4-ol

Dodavatel: Apollo Scientific
Varování

PC28240-1G PC28240-250MG
APOSPC28240-1GEA 6850 CZK
APOSPC28240-1G APOSPC28240-250MG
6-Fluoro-8-(trifluoromethyl)quinolin-4-ol
6-Fluoro-8-(trifluoromethyl)quinolin-4-ol
Vzorec: C₁₀H₅F₄NO
MW: 231,146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28010367
CAS číslo: 1065092-53-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR